missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor N100512KG

 View more versions of this product

Catalog No. 18611901


Explore available promotions

Description

Description

Specifications

Specifications

91-20-3
Poly Pail
0.99
1
12 kg
Reagent

Chemical Identifiers

91-20-3
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.