Learn More
Naphthenic acids, pract., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 415281000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Naphthenic acids | |
230 to 260mg KOH/g | |
0.9100g/mL | |
149°C | |
Glass bottle | |
100 g | |
Solubility in water: insoluble | |
Liquid |
pract. | |
1338-24-5 | |
140.0°C to 200.0°C | |
Authentic | |
MFCD00147696 | |
0.91 | |
5 stokes max. (25°C) |
Chemical Identifiers
1338-24-5 | |
MFCD00147696 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 215-662-8
RTECSNumber : QK8750000
TSCA : TSCA
RUO – Research Use Only