missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
α-Naphthyl phosphate, 99.8%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210025701
Specifications
α-Naphthyl phosphate | |
White | |
C10H7Na2O4P | |
1 g | |
QYURIFWAOPAPAJ-UHFFFAOYSA-L | |
disodium naphthalen-1-yl phosphate | |
75132 | |
Powder |
2183-17-7 | |
0.998 | |
MFCD00041007 | |
disodium 1-naphthyl phosphate, sodium naphthalen-1-yl phosphate, disodium naphthyl phosphate, 1-naphthyl phsophoric acid sodium salt, disodium naphthalen-1-yl phosphate, alpha-naphthyl phosphate disodium salt, dipotassium naphthalen-1-yl phosphate, disodium 1-naphthyl phosphate hydrate, sodium alpha-naphthyl phosphate, 1-naphthyl disodium orthophosphate | |
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 | |
268.12 | |
268.1 |
Chemical Identifiers
2183-17-7 | |
268.12 | |
QYURIFWAOPAPAJ-UHFFFAOYSA-L | |
75132 | |
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
C10H7Na2O4P | |
MFCD00041007 | |
disodium 1-naphthyl phosphate, sodium naphthalen-1-yl phosphate, disodium naphthyl phosphate, 1-naphthyl phsophoric acid sodium salt, disodium naphthalen-1-yl phosphate, alpha-naphthyl phosphate disodium salt, dipotassium naphthalen-1-yl phosphate, disodium 1-naphthyl phosphate hydrate, sodium alpha-naphthyl phosphate, 1-naphthyl disodium orthophosphate | |
disodium naphthalen-1-yl phosphate |