missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naringin, Thermo Scientific Chemicals

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activity

Supplier:  Thermo Scientific Chemicals J6316614

 View more versions of this product

Catalog No. AAJ6316614


Explore available promotions

Description

Description

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer′s disease.

Solubility
Soluble in water, alcohol, acetone and warm acetic acid.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Naringin
White to Yellow
MFCD00148888,MFCD00149445,MFCD01461988
102012
4',5,7-Trihydroxyflavanone 7-rhamnoglucoside
DFPMSGMNTNDNHN-ZPHOTFPESA-N
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
74787988
Powder
10236-47-2
C27H32O14
25 g
14,6425
Soluble in water,alcohol,acetone and warm acetic acid.
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
580.54
580.53

Chemical Identifiers

10236-47-2
580.54
DFPMSGMNTNDNHN-ZPHOTFPESA-N
74787988
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
C27H32O14
MFCD00148888,MFCD00149445,MFCD01461988
4',5,7-Trihydroxyflavanone 7-rhamnoglucoside
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 233-566-4

RTECSNumber : QN6340000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only