Fisherbrand™ Narrow Neck Spherical Dewar Flask with Aluminum Housing
Borosilicate glass with silvered walls
Supplier:  Fisherbrand™ FB1019624
Catalog No. FB1019624
Explore available promotions
Description
Description
  • Spherical
  • Narrow Neck
  • Cork Stopper Included
  • For Use with Liquified Gases
  • Lab Grade
Specifications
Specifications
Cork Stopper
4.72 in.
276 mm
Silver
2 L
143 mm
Liquid Nitrogen
200 mm
Aluminum