Nasco Whirl-Pak™ Stand-Up Sample Bags
Stand-Up Bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
Supplier: Nasco B01450
Description
White, write-on strip makes sample identification easier and more convenient
- Paper-like finish virtually eliminates ink smearing
- Surface that will accept a variety of writing instruments
- Sterilized after manufacturing
Specifications
Whirl-Pak™ Stand-Up Bag | |
1.24 L | |
0.102 mm | |
82°C | |
38 x 15 cm (15 x 6 in.) | |
38 cm | |
15 cm |
42 oz. | |
Sterile | |
250/Pk. | |
4 mil | |
15 in. | |
6 in. |