missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Nateglinide | |
White | |
MFCD00875706 | |
nateglinide, starlix, fastic, starsis, senaglinide, trazec, sdz-djn-608, nateglinide inn, unii-41x3pwk4o2, chembl783 | |
CC(C)C1CCC(CC1)C(=O)NC(CC2=CC=CC=C2)C(=O)O | |
317.429 | |
317.43 | |
Crystalline Powder |
105816-04-4 | |
C19H27NO3 | |
1 g | |
OELFLUMRDSZNSF-OFLPRAFFSA-N | |
(2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoic acid | |
5311309 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
105816-04-4 | |
317.429 | |
OELFLUMRDSZNSF-OFLPRAFFSA-N | |
5311309 | |
CC(C)C1CCC(CC1)C(=O)NC(CC2=CC=CC=C2)C(=O)O |
C19H27NO3 | |
MFCD00875706 | |
nateglinide, starlix, fastic, starsis, senaglinide, trazec, sdz-djn-608, nateglinide inn, unii-41x3pwk4o2, chembl783 | |
(2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoic acid |
Safety and Handling
RTECSNumber : SQ7318950
TSCA : No