OHAUS™ Navigator Portable Precision Balance
The Navigator series offers essential weighing applications at a competitive price point.
Supplier: OHAUS™ 30642243
Description
- The Navigator series has advanced weighing technology which offers one-second stabilization for fast and accurate weighing results. This improves operator efficiency, productivity and throughput.
- Navigators have overload protection systems that can withstand loads up to four times the rated capacity safeguards the balance.
Specifications
IEC/EN 61010-1, IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B; Canada ICES-003 Class B, CE, RCM | |
3200 g | |
0.01 g | |
NV3202 | |
230 x 204 x 70 mm |
RS232, USB, or Ethernet (available as accessories) | |
LCD | |
ABS, Stainless Steel | |
2 sec. | |
AC adapter (included) or 4 C batteries (not included) |