missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BTX™ Needle L-Shaped Platinum Electrode
Ultra thin diameter Needle L-Shaped Platinum Electrode for in vivo applications.
Supplier: BTX™ 450509
Description
- Enables pinpoint transfection for greater ease and efficiency in fragile tissue
- L-shaped electrodes available in 3 mm tip length
Specifications
Ex-Vivo or in-Vivo Tissues Gene or Drug Delivery, Nuclear Transfer | |
Platinum | |
Electroporation accessory | |
Needle L-Shaped Platinum Electrode, 3 mm | |
Electrode Only | |
3 mm | |
Non-sterile | |
Digital User |
Gemini X2, ECM 830, ECM 2001+ and Legacy ECM 2001 | |
BTX Electroporation | |
100 VDC | |
L-Shaped | |
0.12 in. | |
10 μsec. to 100 msec. | |
Banana Cables | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.