missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neo-Clear™, (Xylene Substitute), For Microscopy, MilliporeSigma™

For Histoprocessing, Deparaffination and clearing after dehydration in staining process.

Supplier:  Technidata 1098435000

 View more versions of this product

Catalog No. M1098435000


Explore available promotions

Description

Description

Specifications

Specifications

Liquid
0.78 - 0.795 g/3 (20 °C)
6 hPa (20°C)
5 L
64741-65-7
Plastic Bottle
43°C

Chemical Identifiers

64741-65-7
Safety and Handling

Safety and Handling

EINECSNumber : 265-067-2

SDS
Documents

Documents

Promotions

Promotions