missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neo-Clear™, (Xylene Substitute), For Microscopy, MilliporeSigma™
For Histoprocessing, Deparaffination and clearing after dehydration in staining process.
Supplier: Technidata 1098439025
Specifications
Liquid | |
0.78Â -Â 0.795Â g/3 (20Â °C) | |
6 hPa (20°C) | |
25 L |
64741-65-7 | |
Barrel Pe/Met | |
43°C |
Chemical Identifiers
64741-65-7 |
Safety and Handling
EINECSNumber : 265-067-2