missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine hemihydrate, 99+%

CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol

Supplier:  Thermo Scientific Chemicals 153320050

 View more versions of this product

Catalog No. AC153320050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Neocuproine hemihydrate
99.0
158.0°C to 163.0°C
Authentic
Glass bottle
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
15, 6534
Solubility in water: slightly soluble. Other solubilities: 1500g/L methanol (20°C)
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
434.54
217.27
Crystalline Powder
34302-69-7
100.0
White
99% min. (Total base)
C28H26N4O
5 g
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
IEBXFSLFDFHSRD-UHFFFAOYSA-N
bis(2,9-dimethyl-1,10-phenanthroline) hydrate
67652146
99+%

Chemical Identifiers

34302-69-7
434.54
IEBXFSLFDFHSRD-UHFFFAOYSA-N
67652146
C28H26N4O
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and k

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only