missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neocuproine hydrochloride monohydrate, For Spectrophotometric Det. of Cu, ≥99.0%, MilliporeSigma™ Supelco™
$83.41 - $305.95
Chemical Identifiers
MDL Number | MFCD00150062 |
---|---|
Synonym | 2,9-Dimethyl-1,10-phenanthroline hydrochloride |
Chemical Identifiers
MFCD00150062 | |
2,9-Dimethyl-1,10-phenanthroline hydrochloride |
Specifications
303136-82-5 | |
MFCD00150062 | |
2,9-Dimethyl-1,10-phenanthroline hydrochloride | |
262.73 |
C14H12N2 · HCl · H2O | |
1 g | |
Water: 25 mg/mL, clear | |
≥99.0% (AT); ≥99.0% |