missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neodymium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

$99.89 - $352.45

Chemical Identifiers

CAS 14589-38-9
Molecular Formula C15H21NdO6
Molecular Weight (g/mol) 441.57
MDL Number MFCD00013498
InChI Key AZTDRGMPTCCVFW-UHFFFAOYSA-N
Synonym Neodymium acetylacetonate
PubChem CID 122177161
SMILES [Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1318404
View Documents
Thermo Scientific Chemicals
01318404
2 g
Each for $99.89
 
AA1318409
View Documents
Thermo Scientific Chemicals
01318409
10 g
Each for $352.45
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14589-38-9
441.57
AZTDRGMPTCCVFW-UHFFFAOYSA-N
122177161
C15H21NdO6
MFCD00013498
Neodymium acetylacetonate
[Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

150°C to 152°C
Powder
C15H21NdO6
Neodymium acetylacetonate
[Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
441.57
441.57
Odorless
(REO)
14589-38-9
2 g
MFCD00013498
AZTDRGMPTCCVFW-UHFFFAOYSA-N
neodymium(3+) tris(2,4-dioxopentan-3-ide)
122177161
99.9%
Neodymium(III) 2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-631-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only