missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

$252.01 - $1132.23

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4591406
View Documents
Thermo Scientific Chemicals
04591406
5 g
Each for $252.01
 
AA4591414
View Documents
Thermo Scientific Chemicals
04591414
25 g
Each for $1,132.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

7440-00-8
144.24
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
CHEBI:33372
[Nd]

Specifications

Gray to Silver
Nd
UN1383
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
[Nd]
144.24
CHEBI:33372
−200 Mesh
Air and moisture sensitive
Neodymium powder
7440-00-8
MFCD00011130
5 g
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
23934
99.9%
Odorless
Powder
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H250-H260-H252

P210-P235+P410-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q

H228-H315-H319-X

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-109-3

RTECSNumber : QO8575000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only