missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040294DM
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Neodymium rod | |
7440-00-8 | |
Nd | |
100 mm | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
neodymium | |
23934 | |
99.5% | |
Mylar under Argon | |
Cast Rod |
12.7mm (0.5 in.) dia. | |
(metals basis excluding Ta) | |
MFCD00011130 | |
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n | |
[Nd] | |
144.24 | |
CHEBI:33372 | |
Odorless | |
≈0.89g/mm |
Chemical Identifiers
7440-00-8 | |
144.24 | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
23934 | |
neodymium |
Nd | |
MFCD00011130 | |
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n | |
CHEBI:33372 | |
[Nd] |
Safety and Handling
EINECSNumber : 231-109-3
RTECSNumber : QO8575000
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only