missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+)-Neomenthol 96.0+%, TCI America™
$243.87 - $821.21
Chemical Identifiers
CAS | 2216-52-6 |
---|---|
Molecular Formula | C10H20O |
Molecular Weight (g/mol) | 156.269 |
MDL Number | MFCD00062980 |
InChI Key | NOOLISFMXDJSKH-UTLUCORTSA-N |
Synonym | (1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol |
PubChem CID | 439263 |
ChEBI | CHEBI:15402 |
IUPAC Name | (1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
SMILES | CC1CCC(C(C1)O)C(C)C |
Chemical Identifiers
2216-52-6 | |
156.269 | |
NOOLISFMXDJSKH-UTLUCORTSA-N | |
439263 | |
(1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
C10H20O | |
MFCD00062980 | |
(1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol | |
CHEBI:15402 | |
CC1CCC(C(C1)O)C(C)C |
Specifications
2216-52-6 | |
212°C | |
MFCD00062980 | |
(1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol | |
CC1CCC(C(C1)O)C(C)C | |
156.269 | |
CHEBI:15402 | |
≥96.0% (GC) | |
(+)-Neomenthol |
Colorless | |
C10H20O | |
5 mL | |
NOOLISFMXDJSKH-UTLUCORTSA-N | |
(1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
439263 | |
156.27 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-2333
TSCA : Yes