Invitrogen™ Neon™ NxT Electroporation System 1-Channel Pipette Station
The Neon NxT Electroporation System Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1).
Supplier:  Invitrogen™ NEON1PS
Catalog No. NEON1PS
Explore available promotions
Description
Description
The Neon NxT Electroporation System 1-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1S). Each Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S and NEON18SK) comes with a 1-channel pipette station. This 1-channel pipette station is offered separately for those needing an additional pipette station or replacement for their existing pipette station.
Specifications
Specifications
Neon™ NxT 10-μL and 100-μL tips
Transfection
1 station
Electroporation Pipette Station
Neon™ NxT Electroporation System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.