Invitrogen™ Neon™ Transfection System 100 μL Kit DFS Item
Neon™ Transfection System 100 μL Kit
Supplier:  Invitrogen™ MPK10025
 View more versions of this product
Catalog No. MPK10025
Explore available promotions
Description
Description
The Neon™ Transfection System 100 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 100 μL, containing 5 × 105–2 × 106 adherent cells or 1 × 106–5 × 106 suspension cells. Cells that have been transfected using the included 100μL Neon™ Tips are then ready to be washed and plated into a 6-well, 60 mm, or 10 cm culture dish, depending on your protocol.
Specifications
Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA
Electroporation
Transfection
Neon™
25 x 2 Reactions
Room Temperature
Neon™ Transfection System
The Neon™ Transfection System 100 μL Kit comes with 10 mL resuspension buffer R, 10 mL resuspension buffer T, and 75 mL E2 electrolytic buffer in one box and 25 x 100μL reaction tips and 5 electroporation tubes in a separate box. Refer to the Neon™ Transfection System users manual for protocol instructions. Store the buffer box at 4°C and the tips & tubes box at room temperature.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
CMV
Neon™ Transfection System
50 Reactions
Transfection System
Yes
2x10^4 - 6 x10^8 cells; Varies by cell type
100μl
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.