missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neopentyl alcohol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 128252500

 View more versions of this product

Catalog No. AC128252500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Neopentyl alcohol
98.5
0.8100g/mL
37°C
98.5% min. (GC)
C5H12O
MFCD00004682
01, 406
15, 6542
Solubility in water: 3.5% (25 c)
CC(C)(C)CO
88.15
88.15
75-84-3
100.0
113°C to 114°C
Authentic
Glass bottle
(CH3)3CCH2OH
250g
0.81
neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
KPSSIOMAKSHJJG-UHFFFAOYSA-N
2,2-dimethylpropan-1-ol
6404
99%

Chemical Identifiers

75-84-3
88.15
KPSSIOMAKSHJJG-UHFFFAOYSA-N
6404
CC(C)(C)CO
C5H12O
MFCD00004682
neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
2,2-dimethylpropan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-907-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only