Learn More
Neopentyl chloride, 98%, Thermo Scientific Chemicals
$92.96 - $378.01
Chemical Identifiers
CAS | 753-89-9 |
---|---|
Molecular Formula | C5H11Cl |
Molecular Weight (g/mol) | 106.593 |
MDL Number | MFCD00000941 |
InChI Key | JEKYMVBQWWZVHO-UHFFFAOYSA-N |
PubChem CID | 12956 |
IUPAC Name | 1-chloro-2,2-dimethylpropane |
SMILES | CC(C)(C)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0236506
|
Thermo Scientific Chemicals
L0236506 |
5 g |
Each for $92.96
|
|
||||||
AAL0236514
|
Thermo Scientific Chemicals
L0236514 |
25 g |
Each for $378.01
|
|
||||||
Description
Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNeopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and alkali metals.
Chemical Identifiers
753-89-9 | |
106.593 | |
JEKYMVBQWWZVHO-UHFFFAOYSA-N | |
1-chloro-2,2-dimethylpropane |
C5H11Cl | |
MFCD00000941 | |
12956 | |
CC(C)(C)CCl |
Specifications
753-89-9 | |
0.868 | |
−8°C (17°F) | |
C5H11Cl | |
(CH3)3CCH2Cl | |
5 g | |
1730985 | |
Miscible with organic solvents. Immiscible with water. | |
CC(C)(C)CCl | |
106.593 | |
106.6 | |
Neopentyl chloride |
-20°C | |
83°C to 84°C | |
Sweet | |
1.4039 | |
MFCD00000941 | |
UN1107 | |
neopentyl chloride, propane, 1-chloro-2,2-dimethyl, neo-pentylchloride, t-c4h9ch2cl, 2,2-dimethylpropyl chloride, acmc-1bc1q, neopentyl chloride 5g, propane,1-chloro-2,2-dimethyl, 1-chloro-2,2-dimethylpropane | |
JEKYMVBQWWZVHO-UHFFFAOYSA-N | |
1-chloro-2,2-dimethylpropane | |
12956 | |
98% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
EINECSNumber : 212-040-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only