missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neopentyl Glycol Diglycidyl Ether 40.0+%, TCI America™
$65.17 - $314.03
Chemical Identifiers
CAS | 17557-23-2 |
---|---|
Molecular Formula | C11H20O4 |
Molecular Weight (g/mol) | 216.277 |
MDL Number | MFCD00040687 |
InChI Key | KUAUJXBLDYVELT-UHFFFAOYSA-N |
Synonym | Diglycidyloxyneopentane |
PubChem CID | 28594 |
IUPAC Name | 2-[[2,2-dimethyl-3-(oxiran-2-ylmethoxy)propoxy]methyl]oxirane |
SMILES | CC(C)(COCC1CO1)COCC2CO2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
N044825ML
|
TCI America
N044825ML |
25 mL |
Each for $65.17
|
|
|||||
N0448500ML
|
TCI America
N0448500ML |
500 mL |
Each for $314.03
|
|
|||||
Chemical Identifiers
17557-23-2 | |
216.277 | |
KUAUJXBLDYVELT-UHFFFAOYSA-N | |
28594 | |
CC(C)(COCC1CO1)COCC2CO2 |
C11H20O4 | |
MFCD00040687 | |
Diglycidyloxyneopentane | |
2-[[2,2-dimethyl-3-(oxiran-2-ylmethoxy)propoxy]methyl]oxirane |
Specifications
17557-23-2 | |
C11H20O4 | |
25 mL | |
KUAUJXBLDYVELT-UHFFFAOYSA-N | |
2-[[2,2-dimethyl-3-(oxiran-2-ylmethoxy)propoxy]methyl]oxirane | |
28594 | |
≥40.0% (GC) | |
Neopentyl Glycol Diglycidyl Ether |
Colorless | |
MFCD00040687 | |
Diglycidyloxyneopentane | |
CC(C)(COCC1CO1)COCC2CO2 | |
216.277 | |
216.28 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0396
RTECSNumber : TX3760000
TSCA : Yes