Learn More
Neopentylamine, 97%
CAS: 5813-64-9 | C5H13N | 87.17 g/mol
$262.62 - $262.62
Chemical Identifiers
| CAS | 5813-64-9 |
|---|---|
| Molecular Formula | C5H13N |
| Molecular Weight (g/mol) | 87.17 |
| MDL Number | MFCD00008134 |
| InChI Key | XDIAMRVROCPPBK-UHFFFAOYSA-N |
| Synonym | neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine |
| PubChem CID | 79882 |
| IUPAC Name | 2,2-dimethylpropan-1-amine |
| SMILES | CC(C)(C)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC173690250
|
Thermo Scientific Chemicals
173690250 |
25 mL |
Each for $262.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5813-64-9 | |
| 87.17 | |
| XDIAMRVROCPPBK-UHFFFAOYSA-N | |
| 79882 | |
| CC(C)(C)CN |
| C5H13N | |
| MFCD00008134 | |
| neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
| 2,2-dimethylpropan-1-amine |
Specifications
| 5813-64-9 | |
| 79.0°C to 82.0°C | |
| Authentic | |
| C5H13N | |
| (CH3)3CCH2NH2 | |
| 25 mL | |
| 0.74 | |
| Solubility in water: soluble. | |
| CC(C)(C)CN | |
| 87.17 | |
| 87.16 | |
| Neopentylamine |
| 0.7400g/mL | |
| −13°C | |
| 96% min. (GC) | |
| 1.403 | |
| MFCD00008134 | |
| 04,188 | |
| neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
| XDIAMRVROCPPBK-UHFFFAOYSA-N | |
| 2,2-dimethylpropan-1-amine | |
| 79882 | |
| 97% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 227-378-1
RUO – Research Use Only