missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ NERL™ Blood Bank Saline
Enhance weak agglutination reactions with this saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood.
$125.03 - $152.53
Specifications
Certifications/Compliance | Certificates of Analysis are available online for this product |
---|---|
Content And Storage | 15°C to 30°C |
Description | Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated. |
Stability | 12 Months |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
23-332-408
|
Thermo Scientific™
8504 |
Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated. | 1 x 20 L |
Each for $152.53
|
|
||||
23-249-605
|
Thermo Scientific™
8505 |
Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated. | 1 x 10 L |
Each for $125.03
|
|
||||
Description
Enhance weak agglutination reactions with Thermo Scientific™ NERL™ Blood Bank Saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood. Saline is available in Cubitainer™ containers with spigots for ease of use.
- Phosphate buffered
- Preservative-free
- For in vitro diagnostics and laboratory use only, in immunohematological procedure or as a general laboratory diluent when “normal” or isotonic saline is indicated
- Not for infusion, injection or irrigation
Specifications
Certificates of Analysis are available online for this product | |
15°C to 30°C | |
Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated. | |
12 Months |
General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.