missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ NERL™ High Purity Water

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.

$54.75 - $100.93

Specifications

pH 5 to 7
Grade NERL
Quantity 1 gal.

Alert:

  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
23-249-589
View Documents
Thermo Scientific™
9801
1 gal.
Each for $54.75
Case of 4 Each for $219.00
 
23-249-590
View Documents
Thermo Scientific™
9805
5 gal.
Each for $100.93
 
Description

Description

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.

Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.

Contains no preservatives

Ordering Information:

Certificate of analysis available for each case

Specifications

Specifications

5 to 7
NERL
1 gal.
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Recommended Storage : 15° to 30°C

SDS
missing translation for 'documents'

missing translation for 'documents'

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.