missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ NERL™ Reagent Grade Water
Improve control in critical diagnostic procedures by using this reagent grade water in place of deionized or distilled water.
$106.40 - $221.75
Specifications
Purity Grade Notes | Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online. |
---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
23-249-580
![]() |
Thermo Scientific™
98003 |
32 oz. |
Case of 6 Each for $141.14
|
![]() |
|||||
23-249-579
|
Thermo Scientific™
98001 |
1 gal. |
Case of 4 Each for $221.75
|
|
|||||
23-249-581
|
Thermo Scientific™
98005 |
5 gal. |
Each for $106.40
|
|
|||||
Description
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ Reagent Grade Water in place of deionized or distilled water. This water meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.
- Ideal for reconstituting chemistry or coagulation products, preparing analytical standards or rinsing delicate electrodes
- Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron
- Essentially free from organic and inorganic particulate and soluble contaminants
- Contains no preservatives
- Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water
- Certificate of Analysis is available online
Specifications
Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online. |
General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.