missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-pNP 93.0+%, TCI America™
Supplier: TCI America N08562MG
Specifications
| Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-pNP | |
| White-Yellow | |
| 2 mg | |
| CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)NC(=O)C)OC4=CC=C(C=C4)[N+](=O)[O-])CO)O)O)O)O | |
| 795.701 | |
| 795.70 | |
| Crystalline Powder |
| 501427-93-6 | |
| C31H45N3O21 | |
| TVDCKIMQUNLKLE-SJBDCPKFSA-N | |
| 5-acetamido-2-[[6-[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid | |
| 133612471 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 501427-93-6 | |
| 795.701 | |
| 133612471 | |
| CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)NC(=O)C)OC4=CC=C(C=C4)[N+](=O)[O-])CO)O)O)O)O |
| C31H45N3O21 | |
| TVDCKIMQUNLKLE-SJBDCPKFSA-N | |
| 5-acetamido-2-[[6-[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer