Agilent BioTek™ Neurite Outgrowth Module for Gen5 Software
For use with Gen5 Image Prime Software ≥ v3.16 (part number GEN5IPRIME), purchased separately.
Supplier: Agilent BioTek™ GEN5NEURITE
Description
For use with Gen5 Image Prime Software ≥ v3.16 (part number GEN5IPRIME), purchased separately.
- The Agilent BioTek Gen5 neurite outgrowth module accurately quantifies neuronal cell metrics and provides masking options including soma and neurite masks, along with skeletonized neurite masks.
- The Gen5 neurite outgrowth module accurately detects neuronal outgrowth in kinetically monitored, unlabeled live cells.
Module, Software, Neurite, Outgrowth module
Specifications
Gen5 Image Prime | |
Software |