Gibco™ Neurobasal™ Medium DFS Item
Used in the growth of neuronal cells from hippocampus, cortex and otregions of the brain. Gibco™ Neurobasal™ Medium is a basal medium that meets the special cell culture requirements of pre-natal and embryonic neuronal cells when used with Gibco™ B-27™ Supplement.
Supplier:  Gibco™ 21103049
Catalog No. 21103049
Explore available promotions
Description
Description
  • Allows for both long and short term maintenance of homogeneous populations of neuronal cells without the need of an astrocyte feeder layer
  • Supplemented with 0.5mM L-glutamine or GlutaMAX™-I Supplement and either a serum-free supplement (such as B-27™ Supplement or N-2 Supplement), or with serum
  • Tested for pH, osmolality, endotoxin, and the absence of bacterial and fungal contamination
Order Info
Room Temperature
Specifications
Specifications
Store in refrigerator (2–8°C) and protect from light.
Serum-free
Liquid
Phenol Red
500 mL
Endotoxin, Performance, Sterility, Osmolality, pH
Neural Cells
Neuronal Cell Culture
Neurobasal Medium
Neurobasal™
Ambient
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.