Learn More
Scigen Neutralex™ Neutra-Form™ Formalin Spill Control
Neutra-Form™ Spill Control granules are designed to neutralize hazardous aldehydes, aldehyde vapors, and formalin spills, from 500 mL to 1 gallon. Neutralex™ products are certified by the California Environmental Protection Agency.
Supplier: Scigen SGN4304
Description
Neutra-Form™ is a granule that controls, absorbs, neutralizes accidental spills of aldehyde and formalin. In the event of a spill, simply surround the spill with Neutra-Form granules™ — allowing them to absorb the liquid.
The treated granules provide maximum containment for large and small spills neutralizing liquid while diminishing harmful vapors. Once the Neutra-Form™ granules have absorbed the spill, simply sweep up the residue and place it in a proper disposal container.
Neutra-Wipes™ are designed specifically for use with other Neutralex™ products for maximum effectiveness in treating formalin and aldehyde waste.
- Each container treats and absorbs up to 500 mL of 10% formalin
- Manufactured using ISO 13485 quality system
Specifications
White, Gray | |
3 lb. | |
Neutra-Form™ | |
2/Cs. | |
3.3 kg |
Instantaneously neutralex formalin spills | |
1.4 kg | |
Formalin, glutaraldehyde, paraldehyde and OPA large spills | |
7.3 lb. |