Eppendorf™ Scientific Shaker Utility Trays
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
$1577.63 - $1838.05
Specifications
Includes Non-skid Rubber surface
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Platform Width (Metric) Platform Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Platform Width (Metric) Platform Product Type Price Quantity  
14-278-129
View Documents
Eppendorf™
M11949910
Innova 2100, Excella E24/E24R, I-24/!-24R 46 cm 46 cm Utility Tray
Each for $1,577.63
05414367
View Documents
Eppendorf™
AG00*
Excella E5/E10, I-2500, I-2500KC 76 cm 46 cm Utility Tray (for low speed applications)
Each for $1,838.05
Description
Description
  • For shaking vessels without fasteners
  • Trays have nonskid rubber mat
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Specifications
Non-skid Rubber surface
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'