Bullard™ New EVA PAPR Hood Kit
EVASTLN2K features the Sightline hood, offering 320 degree onubstructed vision and unparalled comfort. PAFLEXBT breathing tube adds to operator experience and reduces SKUs. This new hood is paired with the reliable EVA PAPR unit which provides up to 8.5 CFM airflow.
Supplier:  Bullard™ EVASTLN2K
Catalog No. 17-623-658
Explore available promotions
Description
Description
This kit features the EVA blower unit and new EVA PAPR hood, which has a two speed blower and can provide up to 8.5 CFM airflow to the operator.
  • Dual alarms and visual fuel gauge are just some of the blower features.
  • The belt is comfortable and easy to clean.
  • Bullard's new Sightline hood sets the industry standard with maximum vision, beveled bibs, three point air distibution and innovative drawsting/clamp combination to maximixe fit and comfort.
  • The PAFLEXBT contours to operators body, and prevents and bowing effect.
Specifications
Specifications
Black, White
Available with a High Efficiency HEPA filter, Organic Vapor Cartridge/Filter Combo, or Acid Gas Cartridge/Filter Combo
PAPR Kit
One Size