missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

New Fuchsin, Thermo Scientific Chemicals

$90.23 - $302.74

Chemical Identifiers

CAS 3248-91-7
Molecular Formula C22H24ClN3
Molecular Weight (g/mol) 365.91
MDL Number MFCD00012567
InChI Key ORDGVBRMUJCHEO-UHFFFAOYSA-M
Synonym Basic Violet 2, C.I. 42520, Magenta III
PubChem CID 14454445
ChEBI CHEBI:87671
IUPAC Name 4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride
SMILES [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC212140250
SDS View Documents
Thermo Scientific Chemicals
212140250
25 g Glass bottle
Each for $90.23
 
AC212141000
SDS View Documents
Thermo Scientific Chemicals
212141000
100 g Glass bottle
Each for $302.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3248-91-7
365.91
ORDGVBRMUJCHEO-UHFFFAOYSA-M
14454445
4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride
C22H24ClN3
MFCD00012567
Basic Violet 2, C.I. 42520, Magenta III
CHEBI:87671
[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1

Specifications

1500 (at lambda min.)
1.1 to 1.4
MFCD00012567
ORDGVBRMUJCHEO-UHFFFAOYSA-M
[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1
365.91
CHEBI:87671
365.89
Green
Crystalline Powder or Crystals
3248-91-7
C22H24ClN3
Basic Violet 2, C.I. 42520, Magenta III
Authentic
4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride
14454445
550 to 554nm (in 50% ethanol)
Glass bottle
25 g
New Fuchsin
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 221-831-7

RTECSNumber : CX9825000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only