missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

New Methylene Blue 'N' Stain, for Reticulocyte Count, Ricca Chemical

For reticulocyte count

$93.91 - $289.03

Chemical Identifiers

CAS 6586-05-6
Molecular Formula C18H22Cl3N3SZn
Molecular Weight (g/mol) 484.19
InChI Key GGKWUSWRGRCLEV-UHFFFAOYSA-K
IUPAC Name zinc(2+) 3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium trichloride
SMILES [Cl-].[Cl-].[Cl-].[Zn++].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5285-4
SDS View Documents
Ricca Chemical Company
52854
120 mL Natural Poly Bottle
Each for $93.91
 
5285-16
View Documents
Ricca Chemical Company
528516
500 mL Natural Poly Bottle
Each for $289.03
 
Description

Description

Specifications

Chemical Identifiers

6586-05-6
484.19
zinc(2+) 3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium trichloride
C18H22Cl3N3SZn
GGKWUSWRGRCLEV-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[Zn++].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C

Specifications

6586-05-6,6487-48-5,7647-14-5,6586-05-6
99.0,1.39,0.8,0.45
GGKWUSWRGRCLEV-UHFFFAOYSA-K
zinc(2+) 3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium trichloride
Natural Poly Bottle
New Methylene Blue “N”
95.0,1.34,0.76,0.43
C18H22Cl3N3SZn
[Cl-].[Cl-].[Cl-].[Zn++].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C
484.19
120 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'