New Pig Haz-Mat Pads
Stop the spread of acids and bases.
Supplier:  New Pig MAT301
 View more versions of this product
Catalog No. 19-039-154
Explore available promotions
Description
Description
Stop the spread of acids and bases.
  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste
Specifications
Specifications
Pink
0.83 L
Hazmat Absorbent Pad
20 L x 15 in. W (50.8 x 38 cm)
20 x 15 in. (50.8 x 38 cm)
50.8 cm
38 cm
7.7 kg
28.16 oz.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
Heavyweight
20 in.
15 in.
17 lb.