New Pig Haz-Mat Pads
Stop the spread of acids and bases.
Supplier:  New Pig MAT309
 View more versions of this product
Catalog No. 19-039-155
Explore available promotions
Description
Description
Stop the spread of acids and bases.
  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste
Specifications
Specifications
Pink
152.2 L
Hazmat Absorbent Pad
150 ft. L x 30 in. W (45.7 m x 76.2 cm)
150 ft. x 30 in. (45.7 m x 76.2 cm)
45.7 m
76.2 cm
13.6 kg
40.2 gal.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
Heavyweight
150 ft.
30 in.
30 lb.