New Pig Haz-Mat Pads
Stop the spread of acids and bases.
Supplier: New Pig MAT302
Description
Stop the spread of acids and bases.
- Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
- Excellent for quickly diking and stopping the spread of chemical spills
- Absorb and retain most acids, bases and unknown liquids
- Can be incinerated after use to reduce waste
Specifications
Pink | |
0.416 L | |
Hazmat Absorbent Pad | |
20 L x 15 in. W (50.8 x 38 cm) | |
20 in. | |
15 in. | |
17 lb. |
14.08 oz. | |
Polypropylene | |
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives | |
20 x 15 in. (50.8 x 38 cm) | |
50.8 cm | |
38 cm | |
7.7 kg |