Rubbermaid™ HYGEN™ High-Performance Damp Mops
Nylon/polyester blend microfiber provides optimal damp mopping performance
Supplier:  Rubbermaid™ Q41000BL
 View more versions of this product
Catalog No. 19-130-0862
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Dense 3 - 5 micrometer quality fiber
  • Advanced hook-and-loop backing
  • Double finished edges
  • Color coded products help reduce cross contamination by differentiating area of use
  • Available in green, blue and red colors
Specifications
Specifications
Damp Mop
18.5 in.
5.5 in.
To Remove Dust, Dirt, Microbes
Blue
0.5 in.
3 to 5 μm, Hook, Loop Backing, Double Finish Edges
Nylon, Polyester Blend Microfiber