Celestron™ Celestron NexYZ 3-Axis Universal Smartphone Adapter
Celestron NexYZ 3-Axis Universal Smartphone Adapter connects a smartphone to a telescope, spotting scope, binocular, monocular, and microscope to capture images and video through the eyepiece
Supplier:  Celestron™ 81055
Catalog No. S72453
Explore available promotions
Description
Description
Features:
  • Works with a wide range of mobile phone models, including all the latest devices from Samsung, Google, and Apple
  • Features three directional knobs that perfectly align the phone camera with the eyepiece in seconds
  • Stands up to rigorous field use thanks to its robust, durable construction with a metal frame and polymer body
  • Fits any eyepiece from 35mm to 60mm in diameter including telescopes with 1.25in. and 2in. eyepieces, spotting scopes, monoculars, and binoculars
  • Attaches to microscopes with the addition of the included adapter ring, taking the usable diameter down to 25mm, the size of a standard microscope eyepiece
  • Strong spring and a threaded twist lock provide a two-level strong and secure grip on the optical instrument’s eyepiece to image with confidence
  • Phone platform is fully adjustable to fit any device, typically with the case on
  • Larger devices like the iPhone 8 Plus and latest Samsung Galaxy devices work as well
  • Secure platform stands up to the weight of heavier devices
  • Spring-loaded clamps make it easy to quickly remove one device and replace it with another
  • Switch image from portrait to landscape by turning the padded eyepiece clamp and readjusting the X, Y, and Z axis
Easy as X-Y-Z
  • Three-axis adjustment gives users the power to place a phone on the platform, center it over the eyepiece with the X and Y knobs, and move up or down over the eyepiece until entire field of view is in the shot
  • Adjustment is precise, quick and easy
  • Switching phones, eyepieces, or optical instruments, only necessitates slight adjustments to re-center the camera over the new eyepiece
Warranty
2-Year Warranty
Specifications
Specifications
Astronomy
NexYZ 3-Axis Universal Smartphone Adapter
Telescope, Spotting scope, Binocular, Monocular, Microscope
Universal Adapter
Metal Frame, Polymer Body
10 oz.
Black
6 x 5.5 x 8 in.
8 in.
6 in.
1
5 in.