missing translation for 'onlineSavingsMsg'
Learn More

Nicergoline

CAS: 27848-84-6 | C24H26BrN3O3 | 484.394 g/mol

$162.25 - $531.36

Identifiants chimiques

CAS 27848-84-6
Molecular Formula C24H26BrN3O3
Molecular Weight (g/mol) 484.394
MDL Number MFCD00869626
InChI Key YSEXMKHXIOCEJA-FVFQAYNVSA-N
Synonym nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit
PubChem CID 34040
IUPAC Name [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
SMILES CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ63295MC
Afficher les documents
Thermo Scientific Chemicals
J63295MC
100 mg
chaque for $162.25
Il en reste null
 
AAJ6329503
Afficher les documents
Thermo Scientific Chemicals
J6329503
1 g
chaque for $531.36
Il en reste null
 
Description

Description

Nicergoline is an ergot derivative and vasodilator which increases blood flow in the brain. Nicergoline inhibits the vasoconstrictor effect of catecholamines by inhibiting the postsynaptic α1-adrenoceptors on vascular smooth muscle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, chloroform, and acetone. Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Spécifications

Identifiants chimiques

27848-84-6
484.394
YSEXMKHXIOCEJA-FVFQAYNVSA-N
34040
CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br
C24H26BrN3O3
MFCD00869626
nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit
[(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate

Spécifications

27848-84-6
C24H26BrN3O3
100 mg
nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit
YSEXMKHXIOCEJA-FVFQAYNVSA-N
[(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
34040
Powder
White
MFCD00869626
14,6496
Soluble in alcohol,chloroform,and acetone. Insoluble in water.
CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br
484.394
484.39
Nicergoline
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 248-694-6

missing translation for 'rtecsNumber' : KE6341000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only