Innovatek Nichrome Innoculating Loops
Innovatek Nichrome Innoculating Loops glide smoothly across the agar surface for evenly isolated colonies
Supplier:  Innovatek 40303730
 View more versions of this product
Catalog No. 361045036
Explore available promotions
Description
Description
  • A wide range of premium quality, accurately sized twisted Nichrome wire loops
  • Nichrome wire offers all the benefits of platinum at much more reasonable cost
  • Accurately formed loops glide smoothly across the agar surface for evenly isolated colonies
  • Exacting manufacturing process ensures a perfectly symmetrical loop with equal dimensions, rather than an uneven oval
  • Calibration gauge checks that inner diameter remains accurate during daily use
  • Consistent and reproducible results
  • Accurately calibrated with Evans Blue dye test
  • Calibration gauge for quality control in the lab
  • Rapid cooling
Specifications
Specifications
Reusable
26 G
Yes
Nichrome Wire
6 in.