Learn More
Nickel(II) bromide, 99%, Thermo Scientific Chemicals
$69.55 - $801.13
Chemical Identifiers
CAS | 13462-88-9 |
---|---|
Molecular Formula | Br2Ni |
Molecular Weight (g/mol) | 218.53 |
MDL Number | MFCD00011141 |
InChI Key | IPLJNQFXJUCRNH-UHFFFAOYSA-L |
Synonym | nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous |
PubChem CID | 278492 |
IUPAC Name | dibromonickel |
SMILES | [Ni](Br)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC203360100
|
Thermo Scientific Chemicals
203360100 |
10 g | Glass bottle |
Each for $69.55
|
|
|||||
AC203360500
|
Thermo Scientific Chemicals
203360500 |
50 g | Glass bottle |
Each for $200.44
|
|
|||||
AC203362500
|
Thermo Scientific Chemicals
203362500 |
250 g | Glass bottle |
Each for $801.13
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13462-88-9 | |
218.53 | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
278492 | |
[Ni](Br)Br |
Br2Ni | |
MFCD00011141 | |
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous | |
dibromonickel |
Specifications
963.0°C | |
Brown | |
100.0 | |
10 g | |
Br2Ni | |
MFCD00011141 | |
15, 6587 | |
Solubility in water: 567g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether | |
[Ni](Br)Br | |
218.53 | |
218.53 | |
Glass bottle |
13462-88-9 | |
98.5 | |
Crystalline Powder and/or Chunks | |
98.5% min. (on dry substance) (Argentometry) | |
NiBr2 | |
04,351 | |
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
dibromonickel | |
278492 | |
99% | |
Nickel(II) bromide |
Safety and Handling
GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May cause allergy or asthma symptoms or brea
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
Avoid release to the environment.
Obtain
GHS Signal Word: Danger
EINECSNumber : 236-665-
RUO – Research Use Only