Learn More
Nickel(II) bromide hydrate, 98%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 214871000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) bromide hydrate | |
98% | |
Orange | |
100.0 | |
Crystalline Powder or Needles | |
97.5% min. (on dry substance) (Argentometry) | |
NiBr2·xH2O | |
04,351 | |
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
dibromonickel;hydrate | |
57377118 | |
98% | |
Glass bottle |
963.0°C | |
207569-11-7,13462-88-9 | |
97.5 | |
5 to 7 (5% aq. soln.) | |
100 g | |
Br2Ni | |
MFCD00149806 | |
10, 6340 | |
Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether | |
[Ni++].[Br-].[Br-] | |
218.50 | |
218.53 | |
Analytical |
Chemical Identifiers
207569-11-7 | |
218.50 | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
57377118 |
Br2Ni | |
MFCD00149806 | |
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci | |
[Ni++].[Br-].[Br-] |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
M
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain speci
GHS Signal Word: Danger
RUO – Research Use Only