Learn More
Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals
$224.62 - $984.32
Chemical Identifiers
CAS | 7789-49-3 |
---|---|
Molecular Formula | Br2H6NiO3 |
Molecular Weight (g/mol) | 272.546 |
MDL Number | MFCD00011141 |
InChI Key | UQPSGBZICXWIAG-UHFFFAOYSA-L |
Synonym | nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at |
PubChem CID | 53471882 |
IUPAC Name | dibromonickel;trihydrate |
SMILES | O.O.O.[Ni](Br)Br |
Description
It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-49-3 | |
272.546 | |
UQPSGBZICXWIAG-UHFFFAOYSA-L | |
53471882 | |
O.O.O.[Ni](Br)Br |
Br2H6NiO3 | |
MFCD00011141 | |
nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at | |
dibromonickel;trihydrate |
Specifications
300°C (in -3H2O) | |
Crystalline Granules | |
Br2H6NiO3 | |
UN3077 | |
nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at | |
UQPSGBZICXWIAG-UHFFFAOYSA-L | |
dibromonickel;trihydrate | |
53471882 | |
98% | |
Hygroscopic |
7789-49-3 | |
50 g | |
MFCD00011141 | |
14,6502 | |
Soluble in water, alcohol | |
O.O.O.[Ni](Br)Br | |
272.546 | |
272.58 (218.53 Anhydrous) | |
Odorless | |
Nickel(II) bromide trihydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350-H360-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 236-665-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only