missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 378350250

Catalog No. AC378350250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
Specifications

Specifications

Nickel(II) chloride
25g
Cl2Ni
MFCD00011142
Solubility in water: slightly soluble
Cl[Ni]Cl
129.6
CHEBI:34887
99.99%
Glass bottle
7718-54-9
44.6 to 46% (Ni) (Complexometry)
NiCl2
15, 6590
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Trace Metal Basis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 231-743-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only