Learn More
Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals
For Metallurgy, Nickel-Plating
Supplier: Thermo Scientific Chemicals 03948018
Description
Applied in metallurgy, nickel-plating industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) cyanide tetrahydrate | |
Green | |
Powder | |
C2H8N2NiO4 | |
nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate | |
VHHVAACTKRUWEK-UHFFFAOYSA-N | |
nickel(2+);dicyanide;tetrahydrate | |
71317429 | |
Light green powder |
200°C-4H2O | |
13477-95-7 | |
50 g | |
MFCD00049490 | |
Insoluble in water. Soluble in ammonium carbonate,ammonia,alkali cyanides | |
[C-]#N.[C-]#N.O.O.O.O.[Ni+2] | |
182.789 | |
182.73 (110.73 Anhydrous) |
Chemical Identifiers
13477-95-7 | |
182.789 | |
VHHVAACTKRUWEK-UHFFFAOYSA-N | |
71317429 | |
[C-]#N.[C-]#N.O.O.O.O.[Ni+2] |
C2H8N2NiO4 | |
MFCD00049490 | |
nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate | |
nickel(2+);dicyanide;tetrahydrate |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350-H372
DOTInformation : DOT Class: 6.1, Packaging Group: II
EINECSNumber : 209-160-8
RTECSNumber : QR6495000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only