missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

$77.77 - $330.16

Chemical Identifiers

CAS 13462-90-3
Molecular Formula I2Ni
Molecular Weight (g/mol) 312.502
MDL Number MFCD00016264
InChI Key BFSQJYRFLQUZKX-UHFFFAOYSA-L
Synonym nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
PubChem CID 26038
IUPAC Name diiodonickel
SMILES [Ni](I)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2289306
View Documents
Thermo Scientific Chemicals
02289306
5 g
Each for $77.77
 
AA2289314
View Documents
Thermo Scientific Chemicals
02289314
25 g
Each for $330.16
 
Description

Description

Nickel(II) iodide serves as a catalyst in carbonylation reactions. It is also used as a reagent in organic synthesis, especially in conjunction with samarium(II) iodide. It plays an important role in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13462-90-3
312.502
BFSQJYRFLQUZKX-UHFFFAOYSA-L
26038
[Ni](I)I
I2Ni
MFCD00016264
nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
diiodonickel

Specifications

797°C
5 g
MFCD00016264
nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
BFSQJYRFLQUZKX-UHFFFAOYSA-L
diiodonickel
26038
99.5%
Hygroscopic; Light sensitive
Nickel(II) iodide, Anhydrous
13462-90-3
I2Ni
UN3077
Soluble in water and alcohols.
[Ni](I)I
312.502
312.51
Odorless
5.834 g/mL
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H341-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 236-666-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only