missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) nitrate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223155000

 View more versions of this product

Catalog No. AC223155000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For analysis
Specifications

Specifications

137.0°C
56.7°C
13478-00-7,13138-45-9
98.5
Crystalline Powder
98.5% min. (Complexometry)
Ni(NO3)2·6H2O
15,6598
Solubility in water: 940g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
290.79
290.8
Analytical
2.05
Nickel(II) nitrate hexahydrate
99%
Blue to Green
100.0
500 g
H12N2NiO12
MFCD00149805
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel(2+);dinitrate;hexahydrate
61630
99%
Plastic bottle

Chemical Identifiers

13478-00-7
290.79
AOPCKOPZYFFEDA-UHFFFAOYSA-N
61630
H12N2NiO12
MFCD00149805
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallow

GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Avoid release to the e

GHS Signal Word: Danger

RTECSNumber : QR7300000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only