missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) octanoate, in mineral spirits (8% Ni)

CAS: 4995-91-9 | C8H16NiO2 | 202.907 g/mol

$101.34 - $101.34

Chemical Identifiers

CAS 4995-91-9
Molecular Formula C8H16NiO2
Molecular Weight (g/mol) 202.907
MDL Number MFCD00064239
InChI Key ANYMYNDNCKURBB-UHFFFAOYSA-N
Synonym nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
PubChem CID 54611835
IUPAC Name nickel;octanoic acid
SMILES CCCCCCCC(=O)O.[Ni]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3947914
View Documents
Thermo Scientific Chemicals
03947914
25 g
Each for $101.34
Only null left
 
Description

Description

Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4995-91-9
202.907
ANYMYNDNCKURBB-UHFFFAOYSA-N
54611835
CCCCCCCC(=O)O.[Ni]
C8H16NiO2
MFCD00064239
nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
nickel;octanoic acid

Specifications

4995-91-9
0.95 g/mL
C8H16NiO2
MFCD00064239
UN1993
Miscible with ethanol,chloroform,acetonitrile,ether,carbon disulfide,petroleum ether and glacial acetic acid. Immiscible with water.
CCCCCCCC(=O)O.[Ni]
202.907
345.1
Liquid
Green
40°C (104°F)
[CH3(CH2)6COO]2Ni
25 g
nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
ANYMYNDNCKURBB-UHFFFAOYSA-N
nickel;octanoic acid
54611835
8% Ni
Nickel(II) octanoate, In mineral spirits
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H340-H350-H360-H372-H304-H226-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
May cause an allergic skin reaction.

P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280g-P281-P285-P301+P310-P303+P361+P353-P304+P341-P308+P313-P331-P333+P313-P342+P311-P363-P370+P378q-P501c

H226-H304-H317-H334-H340-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only