Learn More
Nickel(II) oxalate dihydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03945418
Description
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) oxalate dihydrate | |
Odorless | |
MFCD00167155 | |
UN3077 | |
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate | |
VNYOIRCILMCTHO-UHFFFAOYSA-L | |
nickel(2+);oxalate;dihydrate | |
516789 | |
Powder |
6018-94-6 | |
C2H4NiO6 | |
50 g | |
14,6514 | |
Soluble in acids; Insoluble in water | |
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2] | |
182.741 | |
182.76 (146.73 Anhydrous) |
Chemical Identifiers
6018-94-6 | |
182.741 | |
VNYOIRCILMCTHO-UHFFFAOYSA-L | |
516789 | |
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2] |
C2H4NiO6 | |
MFCD00167155 | |
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate | |
nickel(2+);oxalate;dihydrate |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P312-P330-P342+P311-P363-P501c
H302+H312-H334-H350-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 208-933-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only